Search Thermo Fisher Scientific
It is used as an active pharmaceutical intermediate and in the synthesis of nε-(7-diethylaminocoumarin-3-carboxyl)- and nε-(7-methoxycoumarin-3-carboxyl)-l-fmoc lysine as tools for protease cleavage detection by fluorescence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.