Thermo Scientific Chemicals

N-Fmoc-1-trityl-L-histidine, 98%, Thermo Scientific Chemicals

Catalog number: H59711.06
5 g, Each
Thermo Scientific Chemicals

N-Fmoc-1-trityl-L-histidine, 98%, Thermo Scientific Chemicals

Catalog number: H59711.06
5 g, Each
Quantity
Catalog number: H59711.06
also known as H59711-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109425-51-6
IUPAC Name
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[1-(triphenylmethyl)-1H-imidazol-4-yl]propanoic acid
Molecular Formula
C40H33N3O4
InChI Key
XXMYDXUIZKNHDT-UHFFFAOYNA-N
SMILES
OC(=O)C(CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Assay (HPLC)
97.5% min

Description

It is used in the synthesis and application of Fmoc-His(3-Bum)-OH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and application of Fmoc-His(3-Bum)-OH.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from heat. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text