Thermo Scientific Chemicals

Methyl hydrogen succinate, 95%, Thermo Scientific Chemicals

Catalog number: H59722.22
100 g, Each
Thermo Scientific Chemicals

Methyl hydrogen succinate, 95%, Thermo Scientific Chemicals

Catalog number: H59722.22
100 g, Each
Quantity
Catalog number: H59722.22
also known as H59722-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3878-55-5
IUPAC Name
4-methoxy-4-oxobutanoic acid
Molecular Formula
C5H8O4
InChI Key
JDRMYOQETPMYQX-UHFFFAOYSA-N
SMILES
COC(=O)CCC(O)=O
Assay from Supplier's CofA
≥94.0%
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Melting Point
56.0-61.0°C

Description

It could react with 1,2,3-trimethoxy-benzene, and obtain the 4-oxo-4-(2,3,4-trimethoxy-phenyl)-butyric acid methyl ester. This reaction needs the reagent of PPA. Butanedioic Acid 1-Methyl Ester is an intermediate in the synthesis of agonist to both GHB and GABAB neural receptors, exhibiting neurotransmitter-like effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It could react with 1,2,3-trimethoxy-benzene, and obtain the 4-oxo-4-(2,3,4-trimethoxy-phenyl)-butyric acid methyl ester. This reaction needs the reagent of PPA. Butanedioic Acid 1-Methyl Ester is an intermediate in the synthesis of agonist to both GHB and GABAB neural receptors, exhibiting neurotransmitter-like effects.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text