It is used in electroplating industry. Tin methanesulfonate is an aqueous solution used in the metal surface treatment. Mainly as a replacement for Tin sulfate where high speed deposits are needed (reel-to-reel).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in electroplating industry. Tin methanesulfonate is an aqueous solution used in the metal surface treatment. Mainly as a replacement for Tin sulfate where high speed deposits are needed (reel-to-reel).
Solubility
Not miscible or difficult to mix in water.
Notes
Air sensitive. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- Kok Kee Yang, et al. Diffusion Coefficient of Tin(II) Methanesulfonate in Ionic Liquid and Methane Sulfonic Acid (MSA) Solvent.Metall. Mater. Trans. B.,2011,42(6), 1274-1279.
- Algirdas Vaškelis, et al. Silver nanostructured catalyst for modification of dielectrics surface.Electrochim. Acta.,2005,50(23), 4586-4591.