Thermo Scientific Chemicals

L-Aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H59810.06
5 g, Each
Thermo Scientific Chemicals

L-Aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H59810.06
5 g, Each
Quantity
Catalog number: H59810.06
also known as H59810-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3057-74-7
IUPAC Name
2-amino-4-(tert-butoxy)-4-oxobutanoic acid
Molecular Formula
C8H15NO4
InChI Key
MXWMFBYWXMXRPD-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)CC(N)C(O)=O
Assay (HPLC)
>97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text