3-(Boc-aminomethyl)azetidine hydrochloride, 95%, Thermo Scientific Chemicals
3-(Boc-aminomethyl)azetidine hydrochloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Boc-aminomethyl)azetidine hydrochloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
250 mg
Catalog number H59816.ME
also known as H59816-ME
Price (USD)/ Each
232.00
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
232.00
Add to cart
3-(Boc-aminomethyl)azetidine hydrochloride, 95%, Thermo Scientific Chemicals
Catalog numberH59816.ME
Price (USD)/ Each
232.00
-
Add to cart
Specifications
Chemical Name or Material3-(Boc-aminomethyl)azetidine hydrochloride
CAS91188-15-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Kondo, T., et al. Design and synthesis of new potent dipeptidyl peptidase IV inhibitors with enhanced ex vivo duration. Bioorganic & Medicinal Chemistry. 2007, 15 (7), 2631-2650.
  2. F.-H. Tsai; C. G. Overberger; and; R. Zand. Synthesis and peptide bond orientation in tetrapeptides containing L-azetidine-2-carboxylic acid and L-proline. Biopolymers, 1990, 30 (11-12), 1039-1049.