Thermo Scientific Chemicals

N-Boc-L-glutamic acid 1-tert-butyl ester, 99%, Thermo Scientific Chemicals

Catalog number: H59842.03
1 g, Each
Thermo Scientific Chemicals

N-Boc-L-glutamic acid 1-tert-butyl ester, 99%, Thermo Scientific Chemicals

Catalog number: H59842.03
1 g, Each
Quantity
Catalog number: H59842.03
also known as H59842-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24277-39-2
Assay (HPLC)
>98.5%

Description

Boc-Glu-OtBu is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make unique peptides containing glutamate tert-butyl ester residues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boc-Glu-OtBu is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make unique peptides containing glutamate tert-butyl ester residues.

Solubility
Soluble in dimethyl formamide.

Notes
Recommended storage temperature: -20°C. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text