Boc-Glu-OtBu is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make unique peptides containing glutamate tert-butyl ester residues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Boc-Glu-OtBu is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make unique peptides containing glutamate tert-butyl ester residues.
Solubility
Soluble in dimethyl formamide.
Notes
Recommended storage temperature: -20°C. Store away from oxidizing agents.
RUO – Research Use Only
Karnail S. Atwal.;George C. Rovnyak.; Joseph Schwartz.; Suzanne Moreland.; Anders Hedberg.; Jack Z. Gougoutas.; Mary F. Malley.; David M. Floyd. Dihydropyrimidine calcium channel blockers: 2-heterosubstituted 4-aryl-1,4-dihydro-6-methyl-5-pyrimidinecarboxylic acid esters as potent mimics of dihydropyridines. J. Med. Chem. 1990, 33 (5), 1510-1515.