Thermo Scientific Chemicals

L-Isoleucine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H59843.14
25 g, Each
Thermo Scientific Chemicals

L-Isoleucine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H59843.14
25 g, Each
Quantity
Catalog number: H59843.14
also known as H59843-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69320-89-4
IUPAC Name
tert-butyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
Molecular Formula
C10H22ClNO2
InChI Key
IFRYMHOZFAPYPJ-HAGKNZRZNA-N
SMILES
Cl.CC[C@H](C)[C@H](N)C(=O)OC(C)(C)C
Proton NMR
>97.5%

Description

It is used as a pharmaceutical industry. t is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical industry. t is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuffs.

Solubility
Soluble in 50 mg/ml methanol.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text