2-Amino-4,5-dimethylthiazole hydrochloride, 97%, Thermo Scientific Chemicals
2-Amino-4,5-dimethylthiazole hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-4,5-dimethylthiazole hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number H59878.09
also known as H59878-09
Price (USD)/ Each
82.90
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
82.90
Add to cart
2-Amino-4,5-dimethylthiazole hydrochloride, 97%, Thermo Scientific Chemicals
Catalog numberH59878.09
Price (USD)/ Each
82.90
-
Add to cart
Chemical Identifiers
CAS71574-33-9
IUPAC Namedimethyl-1,3-thiazol-2-amine hydrochloride
Molecular FormulaC5H9ClN2S
InChI KeyXYTUTNQRQLAZLK-UHFFFAOYSA-N
SMILESCl.CC1=C(C)N=C(N)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only