Thermo Scientific Chemicals

N-Boc-L-serine methyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H59889.09
10 g, Each
Thermo Scientific Chemicals

N-Boc-L-serine methyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H59889.09
10 g, Each
Quantity
Catalog number: H59889.09
also known as H59889-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2766-43-0
IUPAC Name
methyl 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoate
Molecular Formula
C9H17NO5
InChI Key
SANNKFASHWONFD-UHFFFAOYNA-N
SMILES
COC(=O)C(CO)NC(=O)OC(C)(C)C
Assay (HPLC)
>94.0%

Description

N-tert-Butoxycarbonyl-L-serine Methyl Ester is an amino acid derivative used in the preparation of Tn antigen, L-glutamic acid p-nitroanilide analogs and new biomedical polymers having Serine and Threonine side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-tert-Butoxycarbonyl-L-serine Methyl Ester is an amino acid derivative used in the preparation of Tn antigen, L-glutamic acid p-nitroanilide analogs and new biomedical polymers having Serine and Threonine side groups.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text