Thermo Scientific Chemicals

1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%, Thermo Scientific Chemicals

Catalog number: H59893.18
50 g, Each
Thermo Scientific Chemicals

1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%, Thermo Scientific Chemicals

Catalog number: H59893.18
50 g, Each
Quantity
Catalog number: H59893.18
also known as H59893-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
663199-28-8
IUPAC Name
1-butyl-3-methyl-1H-imidazol-3-ium dibutyl phosphate
Molecular Formula
C16H33N2O4P
InChI Key
NTXQJRGQUZXUMU-UHFFFAOYSA-M
SMILES
CCCCN1C=C[N+](C)=C1.CCCCOP([O-])(=O)OCCCC
Proton NMR
>95.0%

Description

1-Butyl-3-methylimidazolium dibutyl phosphate is employed as an imidazolium compound salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butyl-3-methylimidazolium dibutyl phosphate is employed as an imidazolium compound salt.

Solubility
Insoluble in water.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text