Thermo Scientific Chemicals

3-Methyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H59899.03
1 g, Each
Thermo Scientific Chemicals

3-Methyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H59899.03
1 g, Each
Quantity
Catalog number: H59899.03
also known as H59899-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
402-61-9
IUPAC Name
5-methyl-1H-pyrazole-3-carboxylic acid
Molecular Formula
C5H6N2O2
InChI Key
WSMQKESQZFQMFW-UHFFFAOYSA-N
SMILES
CC1=CC(=NN1)C(O)=O
Assay (HPLC)
>96.0%

Description

3-Methylpyrazole-5-carboxylic acid is a potent and selective D-amino acid oxidase (DAO) (also known as DAAO, DAMOX and OXDA) inhibitor that protects DAO cells from oxidative stress induced by D-Serine. 3-Methylpyrazole-5-carboxylic acid specifically prevents formalin-induced tonic pain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylpyrazole-5-carboxylic acid is a potent and selective D-amino acid oxidase (DAO) (also known as DAAO, DAMOX and OXDA) inhibitor that protects DAO cells from oxidative stress induced by D-Serine. 3-Methylpyrazole-5-carboxylic acid specifically prevents formalin-induced tonic pain.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text