Thermo Scientific Chemicals

4-Bromo-3,5-difluorobenzoic acid, 96%, Thermo Scientific Chemicals

Catalog number: H59904.03
1 g, Each
Thermo Scientific Chemicals

4-Bromo-3,5-difluorobenzoic acid, 96%, Thermo Scientific Chemicals

Catalog number: H59904.03
1 g, Each
Quantity
Catalog number: H59904.03
also known as H59904-03
Price (USD)
Quantity
-
Have Questions?

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text