4-Chloro-DL-phenylglycine, 98%, Thermo Scientific Chemicals
4-Chloro-DL-phenylglycine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-DL-phenylglycine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number H59921.22
also known as H59921-22
Price (USD)/ Each
387.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
387.00
Add to cart
4-Chloro-DL-phenylglycine, 98%, Thermo Scientific Chemicals
Catalog numberH59921.22
Price (USD)/ Each
387.00
-
Add to cart
Chemical Identifiers
CAS6212-33-5
IUPAC Name(2R)-2-azaniumyl-2-(4-chlorophenyl)acetate
Molecular FormulaC8H8ClNO2
InChI KeyQGJGBYXRJVIYGA-SSDOTTSWSA-N
SMILES[NH3+][C@@H](C([O-])=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 0.1mol/L NaOH (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only