Thermo Scientific Chemicals

Azetidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H59935.03
1 g, Each
Thermo Scientific Chemicals

Azetidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H59935.03
1 g, Each
Quantity
Catalog number: H59935.03
also known as H59935-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36520-39-5
IUPAC Name
azetidine hydrochloride
Molecular Formula
C3H8ClN
InChI Key
HGQULGDOROIPJN-UHFFFAOYSA-N
SMILES
Cl.C1CNC1
Appearance (Color)
White to orange to brown
Form
Powder or crystalline powder or crystals
Assay from Suppliers CofA
≥96.0% (NMR)
Proton NMR
Conforms to structure

Description

Azetidine hydrochloride is a building block for azetidine synthesis and a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azetidine hydrochloride is a building block for azetidine synthesis and a pharmaceutical intermediate.

Solubility
Soluble in water (103.8°C at 760 mmHg).

Notes
Hygroscopic. Reacts with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text