Chemical Identifiers
CAS92954-90-0
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC24H21NO5
InChI KeySWZCTMTWRHEBIN-ANBDAQEENA-N
SMILESOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
View more