Thermo Scientific Chemicals

Iron(III) trifluoromethanesulfonate, tech. 90%, Thermo Scientific Chemicals

Catalog number: H60034.06
5 g, Each
Thermo Scientific Chemicals

Iron(III) trifluoromethanesulfonate, tech. 90%, Thermo Scientific Chemicals

Catalog number: H60034.06
5 g, Each
Quantity
Catalog number: H60034.06
also known as H60034-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63295-48-7
IUPAC Name
iron(3+) tritrifluoromethanesulfonate
Molecular Formula
C3F9FeO9S3
InChI Key
OSHOQERNFGVVRH-UHFFFAOYSA-K
SMILES
[Fe+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Assay (Titration)
>88.0%

Description

Iron(III) triflate is used as an efficient catalyst in the synthesis of 2,3-unsaturated-O-glycosides from 2,4,6-tri-O-acetyl-D-glucal. It is also involved in the synthesis of a variety of beta-enamino ketones and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(III) triflate is used as an efficient catalyst in the synthesis of 2,3-unsaturated-O-glycosides from 2,4,6-tri-O-acetyl-D-glucal. It is also involved in the synthesis of a variety of beta-enamino ketones and esters.

Solubility
Soluble in water, methanol and acetonitrile.

Notes
Moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text