Thermo Scientific Chemicals

Ethyl hydrazinoacetate hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H60042.14
25 g, Each
Thermo Scientific Chemicals

Ethyl hydrazinoacetate hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H60042.14
25 g, Each
Quantity
Catalog number: H60042.14
also known as H60042-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6945-92-2
IUPAC Name
hydrogen ethyl 2-hydrazinylacetate chloride
Molecular Formula
C4H11ClN2O2
InChI Key
HZZRIIPYFPIKHR-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CCOC(=O)CNN
Assay (unspecified)
>96.0%

Description

Ethyl hydrazinoacetate hydrochloride has been used in the preparation of three novel copper(II) complexes as condensation derivatives of 2-pyridinecarboxaldehyde, 2-acetylpyridine and 2-quinolinecarboxaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl hydrazinoacetate hydrochloride has been used in the preparation of three novel copper(II) complexes as condensation derivatives of 2-pyridinecarboxaldehyde, 2-acetylpyridine and 2-quinolinecarboxaldehyde.

Notes
Store in cool dry conditions. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text