Thermo Scientific Chemicals

(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, 95%, Thermo Scientific Chemicals

Catalog number: H60046.03
1 g, Each
Thermo Scientific Chemicals

(-)-1,2-Bis((2R,5R)-2,5-diphenylphospholano)ethane, 95%, Thermo Scientific Chemicals

Catalog number: H60046.03
1 g, Each
Quantity
Catalog number: H60046.03
also known as H60046-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
528565-79-9
IUPAC Name
(2R,5R)-1-{2-[(2R,5R)-2,5-diphenylphospholan-1-yl]ethyl}-2,5-diphenylphospholane
Molecular Formula
C34H36P2
InChI Key
VHHAZLMVLLIMHT-DXCZPEQUNA-N
SMILES
C(CP1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1)P1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Assay from Suppliers CofA
≥94%
Form
Powder or crystals or crystalline powder

Description

It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as ligands like DuPhos and BPE ligands and are highly efficient privileged ligands.

Solubility
Sparingly soluble in water.(3.2E-5 g/L) (25°C

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text