Tris(2,3-dibromopropyl) isocyanurate, 97%, Thermo Scientific Chemicals
Tris(2,3-dibromopropyl) isocyanurate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(2,3-dibromopropyl) isocyanurate, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 g
Catalog number H60056.36
also known as H60056-36
Price (USD)/ Each
232.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
273.00 
Save 40.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
232.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
273.00 
Save 40.35 (15%)
Add to cart
Tris(2,3-dibromopropyl) isocyanurate, 97%, Thermo Scientific Chemicals
Catalog numberH60056.36
Price (USD)/ Each
232.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
273.00 
Save 40.35 (15%)
-
Add to cart
Chemical Identifiers
CAS52434-90-9
IUPAC Nametris(2,3-dibromopropyl)-1,3,5-triazinane-2,4,6-trione
Molecular FormulaC12H15Br6N3O3
InChI KeyNZUPFZNVGSWLQC-UHFFFAOYNA-N
SMILESBrCC(Br)CN1C(=O)N(CC(Br)CBr)C(=O)N(CC(Br)CBr)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
Tris(2,3-dibromopropyl) isocyanurate is a non degradable heterocyclic hexabrominated flame retardant. It inhibited the expression of psaA and psbC that led to the decrease of Nannochloropsis sp.s growth biomass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2,3-dibromopropyl) isocyanurate is a non degradable heterocyclic hexabrominated flame retardant. It inhibited the expression of psaA and psbC that led to the decrease of Nannochloropsis sp.′s growth biomass.

Solubility
Soluble in chloroform.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Ting Ruan.; Yawei Wang.; Chang Wang.; Pu Wang.; Jianjie Fu.; Yongguang Yin.; Guangbo Qu.; Thanh Wang.; Guibin Jiang. Identification and Evaluation of a Novel Heterocyclic Brominated Flame Retardant Tris(2,3-dibromopropyl) Isocyanurate in Environmental Matrices near a Manufacturing Plant in Southern China. Environ. Sci. Technol. 2009, 43 (9),3080-3086.
  2. Nali Zhu.; An Li.; Thanh Wang.; Pu Wang.; Guangbo Qu.; Ting Ruan.; Jianjie Fu.; Bo Yuan.; Lixi Zeng.; Yawei Wang.; Guibin Jiang. Tris(2,3-dibromopropyl) Isocyanurate, Hexabromocyclododecanes, and Polybrominated Diphenyl Ethers in Mollusks from Chinese Bohai Sea.Environ. Sci. Technol. 2012, 46 (13),7174-7181.