(R)-N-Boc-2-hydroxymethylmorpholine, 99%, Thermo Scientific Chemicals
(R)-N-Boc-2-hydroxymethylmorpholine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-N-Boc-2-hydroxymethylmorpholine, 99%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Catalog number H60062.03
also known as H60062-03
Price (USD)/ Each
150.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
150.00
Add to cart
(R)-N-Boc-2-hydroxymethylmorpholine, 99%, Thermo Scientific Chemicals
Catalog numberH60062.03
Price (USD)/ Each
150.00
-
Add to cart
Chemical Identifiers
CAS135065-71-3
IUPAC Nametert-butyl 2-(hydroxymethyl)morpholine-4-carboxylate
Molecular FormulaC10H19NO4
InChI KeyFJYBLMJHXRWDAQ-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCOC(CO)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only