Ii is an intermediate in the metabolism of leucine and also used as flavoring agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ii is an intermediate in the metabolism of leucine and also used as flavoring agents.
Solubility
Soluble in water.
Notes
Refrigerate in tightly sealed containers. Keep away from oxidizing agents.
RUO – Research Use Only
Nobuhiro Sato.; Tomoyuki Matsuura. Studies on pyrazines. Part 32. Synthesis of trisubstituted and tetrasubstituted pyrazines as ant pheromones. J. Chem. Soc. 1996, 1 (19),2345-2350.
Renat Kadyrov.; Thomas H. Riermeier.; Uwe Dingerdissen.; Vitali Tararov.; Armin Börner. The First Highly Enantioselective Homogeneously Catalyzed Asymmetric Reductive Amination: Synthesis of α-N-Benzylamino Acids.J. Org. Chem. 2003, 68 (10), 4067-4070.