Thermo Scientific Chemicals

4-Methyl-2-oxovaleric acid, 94%, Thermo Scientific Chemicals

Catalog number: H60076.06
5 g, Each
Thermo Scientific Chemicals

4-Methyl-2-oxovaleric acid, 94%, Thermo Scientific Chemicals

Catalog number: H60076.06
5 g, Each
Quantity
Catalog number: H60076.06
also known as H60076-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
816-66-0
IUPAC Name
sodium 4-methyl-2-oxopentanoate
Molecular Formula
C6H9NaO3
InChI Key
IXFAZKRLPPMQEO-UHFFFAOYSA-M
SMILES
[Na+].CC(C)CC(=O)C([O-])=O
Assay (GC)
≥92.0%
Appearance (Color)
Clear colorless to yellow
Form
Liquid

Description

Ii is an intermediate in the metabolism of leucine and also used as flavoring agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ii is an intermediate in the metabolism of leucine and also used as flavoring agents.

Solubility
Soluble in water.

Notes
Refrigerate in tightly sealed containers. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text