Chemical Identifiers
CAS11114-20-8
IUPAC Name(2R,3R,4R,5R,6S)-6-{[(1R,3R,4R,5R,8S)-3-{[(2S,3R,4R,5S,6R)-2-{[(1R,3S,4R,5R,8S)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3-hydroxy-6-(hydroxymethyl)-5-(sulfonatooxy)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl sulfate
Molecular FormulaC24H36O25S2
InChI KeyZNOZWUKQPJXOIG-XSBHQQIPSA-L
SMILESOC[C@H]1O[C@@H](O[C@H]2[C@H]3CO[C@@H]2[C@@H](O)[C@@H](O[C@@H]2[C@@H](O)[C@H](O[C@H]4[C@H]5CO[C@@H]4[C@@H](O)[C@@H](O)O5)O[C@H](CO)[C@@H]2OS([O-])(=O)=O)O3)[C@H](O)[C@@H](O)[C@H]1OS([O-])(=O)=O
View more