Thermo Scientific Chemicals

4-Methoxycinnamic acid, 99%, Thermo Scientific Chemicals

Catalog number: H60144.14
25 g, Each
Thermo Scientific Chemicals

4-Methoxycinnamic acid, 99%, Thermo Scientific Chemicals

Catalog number: H60144.14
25 g, Each
Quantity
Catalog number: H60144.14
also known as H60144-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
830-09-1
IUPAC Name
(2E)-3-(4-methoxyphenyl)prop-2-enoic acid
Molecular Formula
C10H10O3
InChI Key
AFDXODALSZRGIH-QPJJXVBHSA-N
SMILES
COC1=CC=C(\C=C\C(O)=O)C=C1
Assay (unspecified)
>98.5%

Description

It exerts anti-hyperglycemic/ hypoglycemic effect by stimulating insulin secretion from pancreas. It could be developed into a new potential therapeutic agent used in type 2 diabetic patients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It exerts anti-hyperglycemic/ hypoglycemic effect by stimulating insulin secretion from pancreas. It could be developed into a new potential therapeutic agent used in type 2 diabetic patients.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool dry conditions. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text