Thermo Scientific Chemicals

Terpineol, mixed isomers, 98%, Thermo Scientific Chemicals

Catalog number: H60146.AP
500 mL, Each
Thermo Scientific Chemicals

Terpineol, mixed isomers, 98%, Thermo Scientific Chemicals

Catalog number: H60146.AP
500 mL, Each
Quantity
Catalog number: H60146.AP
also known as H60146-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8000-41-7
IUPAC Name
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
Molecular Formula
C10H18O
InChI Key
WUOACPNHFRMFPN-UHFFFAOYNA-N
SMILES
CC1=CCC(CC1)C(C)(C)O
Appearance (Color)
White to pale yellow
Form
Liquid or solidified melt
Assay (GC)
>97.5%

Description

Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text