Thermo Scientific Chemicals

Asiatic acid, 97%, Thermo Scientific Chemicals

Catalog number: H60149.ME
500 mg, Each
Thermo Scientific Chemicals

Asiatic acid, 97%, Thermo Scientific Chemicals

Catalog number: H60149.ME
500 mg, Each
Quantity
Catalog number: H60149.ME
also known as H60149-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
464-92-6
IUPAC Name
(1S,2R,4aS,6aS,6bR,8aR,9R,10R,11R,12aR,12bR,14bS)-10,11-dihydroxy-9-(hydroxymethyl)-1,2,6a,6b,9,12a-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid
Molecular Formula
C30H48O5
InChI Key
JXSVIVRDWWRQRT-UYDOISQJSA-N
SMILES
C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)[C@@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C)C(O)=O
Appearance (Color)
White to pale cream
Form
Powder, crystals, crystalline powder and/or chunks
Assay from Supplier's CofA
≥96.0% (HPLC)

Description

Asiatic acid is commonly used in wound healing. Asiatic acid has antioxidant, anti-inflammatory and neuroprotective properties. It is a starting material for asiatic acid derivative synthesis for use as anticancer agents, glycogen phosphorylase inhibitors and and hepatoprotectants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Asiatic acid is commonly used in wound healing. Asiatic acid has antioxidant, anti-inflammatory and neuroprotective properties. It is a starting material for asiatic acid derivative synthesis for use as anticancer agents, glycogen p
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text