2-Piperidinemethanol, 95%, Thermo Scientific Chemicals
2-Piperidinemethanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Piperidinemethanol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number H60178.18
also known as H60178-18
Price (USD)/ Each
300.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
300.00
Add to cart
2-Piperidinemethanol, 95%, Thermo Scientific Chemicals
Catalog numberH60178.18
Price (USD)/ Each
300.00
-
Add to cart
Chemical Identifiers
CAS3433-37-2
IUPAC Name(2R)-2-(hydroxymethyl)piperidin-1-ium
Molecular FormulaC6H14NO
InChI KeyPRAYXGYYVXRDDW-ZCFIWIBFSA-O
SMILESOC[C@H]1CCCC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥94.0%
Appearance (Color)Brown
2-Piperidinemethanol was used in the synthesis of 3-phenyloctahydropyrido[2,1-c][1,4]oxazine hydrochloride and the 10R and 10S diastereomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Piperidinemethanol was used in the synthesis of 3-phenyloctahydropyrido[2,1-c][1,4]oxazine hydrochloride and the 10R and 10S diastereomers.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. T.N.Riley; G.O.Rankin. Synthesis and pharmacological evaluation of some 3-substituted octahydropyrido(2,1-c)(1,4)oxazines. Journal of Medicinal Chemistry. 1976, 19,(2), 334-336.
  2. Masayuki Sato; Charles H.Stammer. Synthesis of N-hydroxythiourea. J. Med. Chem. 1976, 19,(2), 336-337.