It is used as pharmaceutical and fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical and fine chemical intermediate.
Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
Anne Pautard-Cooper.; Slayton A. EvansJr. Mechanistic implications of 1,3,2.lambda.5-dioxaphospholanes in the Mitsunobu reaction. J. Org. Chem. 1989, 54 (10),2485-2488.
Isabel Mathieu-Pelta.; Slayton A. EvansJr. Highly regioselective and stereospecific functionalization of 1,2-propanediol with trimethyl(X)silanes employing the 1,3,2.lambda.5-dioxaphospholane methodology. J. Org. Chem. 1992, 57 (12), 3409-3413.