Appearance (Color)White
Assay from Supplier's CofA≥97.5% (HPLC, US sourced material only)
Water Content (Karl Fischer Titration)≤2.0% (UK sourced material only)
Assay (Titration ex Chloride)≥97.5 to ≤102.5% (Dry wt. basis, UK sourced material only)
CommentUK and US sourced material
View more