Benzyldimethyl-n-dodecylammonium chloride, 98%, Thermo Scientific Chemicals
Benzyldimethyl-n-dodecylammonium chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyldimethyl-n-dodecylammonium chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Catalog number H60196.18
also known as H60196-18
Price (USD)/ Each
387.65
Online exclusive
430.00 
Save 42.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
387.65
Online exclusive
430.00 
Save 42.35 (10%)
Add to cart
Benzyldimethyl-n-dodecylammonium chloride, 98%, Thermo Scientific Chemicals
Catalog numberH60196.18
Price (USD)/ Each
387.65
Online exclusive
430.00 
Save 42.35 (10%)
-
Add to cart
Chemical Identifiers
CAS139-07-1
IUPAC Namebenzyl(dodecyl)dimethylazanium chloride
Molecular FormulaC21H38ClN
InChI KeyJBIROUFYLSSYDX-UHFFFAOYSA-M
SMILES[Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Supplier's CofA≥97.5% (HPLC, US sourced material only)
Water Content (Karl Fischer Titration)≤2.0% (UK sourced material only)
Assay (Titration ex Chloride)≥97.5 to ≤102.5% (Dry wt. basis, UK sourced material only)
CommentUK and US sourced material
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only