6,7-Dihydrobenzo[b]thiophen-4-one, 98%, Thermo Scientific Chemicals
6,7-Dihydrobenzo[b]thiophen-4-one, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6,7-Dihydrobenzo[b]thiophen-4-one, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number H60220.09
also known as H60220-09
Price (USD)/ Each
429.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
429.00
Add to cart
6,7-Dihydrobenzo[b]thiophen-4-one, 98%, Thermo Scientific Chemicals
Catalog numberH60220.09
Price (USD)/ Each
429.00
-
Add to cart
Chemical Identifiers
CAS13414-95-4
IUPAC Name4,5,6,7-tetrahydro-1-benzothiophen-4-one
Molecular FormulaC8H8OS
InChI KeyGJEKNELSXNSYAQ-UHFFFAOYSA-N
SMILESO=C1CCCC2=C1C=CS2
View more
SpecificationsSpecification SheetSpecification Sheet
FormLow-melting solid
Assay (HPLC)>97.5%
Appearance (Color)White to pale brown
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

Solubility
Soluble in chloroform.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Paolo Cozzi.; Umberto Branzoli.; Pier Paolo Lovisolo.; Gaetano Orsini.; Germano Carganico.; Antonio Pillan.; Augusto Chiari. N-Imidazolyl-4-chromanones, N-imidazolyl-1-tetralones, and their alcohols as hypolipemic agents raising high-density lipoproteins. J. Med. Chem. 1986, 29 (3),404-410.
  2. Christoph Jacobs.; Martin Frotscher.; Gerd Dannhardt.; Rolf W. Hartmann. 1-Imidazolyl(alkyl)-Substituted Di- and Tetrahydroquinolines and Analogues: Syntheses and Evaluation of Dual Inhibitors of Thromboxane A2 Synthase and Aromatase. J. Med. Chem. , 2000, 43 (9),1841-1851.