Thermo Scientific Chemicals

2,4-Dibromothiazole, 97%, Thermo Scientific Chemicals

Catalog number: H60230.06
5 g, Each
Thermo Scientific Chemicals

2,4-Dibromothiazole, 97%, Thermo Scientific Chemicals

Catalog number: H60230.06
5 g, Each
Quantity
Catalog number: H60230.06
also known as H60230-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4175-77-3
IUPAC Name
2,4-dibromo-1,3-thiazole
Molecular Formula
C3HBr2NS
InChI Key
MKEJZKKVVUZXIS-UHFFFAOYSA-N
SMILES
BrC1=NC(Br)=CS1
Assay (HPLC)
96.0% min

Description

2,4-Dibromothiazole is applied as a building block in a synthesis of Melithiazole C employing a highly (E)-selective cross-metathesis between the derived 4-vinylthiazole and a dienote side-chain. It is also used as an intermediate in the preparation of various 2,4-disubstituted thiazole derivatives with biologically active properties such as survival motor neuron (SMN) protein modulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromothiazole is applied as a building block in a synthesis of Melithiazole C employing a highly (E)-selective cross-metathesis between the derived 4-vinylthiazole and a dienote side-chain. It is also used as an intermediate in the preparation of various 2,4-disubstituted thiazole derivatives with biologically active properties such as survival motor neuron (SMN) protein modulators.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text