Thermo Scientific Chemicals

5-Methyl-1H-tetrazole, 97%, Thermo Scientific Chemicals

Catalog number: H60248.06
5 g, Each
Thermo Scientific Chemicals

5-Methyl-1H-tetrazole, 97%, Thermo Scientific Chemicals

Catalog number: H60248.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H60248.06
also known as H60248-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4076-36-2
IUPAC Name
5-methyl-2H-1,2,3,4-tetrazole
Molecular Formula
C2H4N4
InChI Key
XZGLNCKSNVGDNX-UHFFFAOYSA-N
SMILES
CC1=NNN=N1
Assay (HPLC)
97%

Description

5-Methyl-1H-tetrazole is used in the preparation of novel antifungal agents based derived from N-iodopropargylazoles and N-triiodoallylazoles. 5-Methyl Tetrazole also in the preparation of novel quinoline derivatives against mycobacterium tuberculosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1H-tetrazole is used in the preparation of novel antifungal agents based derived from N-iodopropargylazoles and N-triiodoallylazoles. 5-Methyl Tetrazole also in the preparation of novel quinoline derivatives against mycobacterium tuberculosis.

Solubility
Soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text