Thermo Scientific Chemicals

Tetradecanedioic acid, 98%, Thermo Scientific Chemicals

Catalog number: H60254.14
25 g, Each
Thermo Scientific Chemicals

Tetradecanedioic acid, 98%, Thermo Scientific Chemicals

Catalog number: H60254.14
25 g, Each
Quantity
Catalog number: H60254.14
also known as H60254-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
821-38-5
IUPAC Name
tetradecanedioic acid
Molecular Formula
C14H26O4
InChI Key
HQHCYKULIHKCEB-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCCCCCCCCC(O)=O
Assay (unspecified)
>97.5%

Description

Tetradecanedioic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetradecanedioic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents amnd bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text