Thermo Scientific Chemicals

O6-Benzylguanine, 98%, Thermo Scientific Chemicals

Catalog number: H60274.MD
250 mg, Each
Thermo Scientific Chemicals

O6-Benzylguanine, 98%, Thermo Scientific Chemicals

Catalog number: H60274.MD
250 mg, Each
Quantity
Catalog number: H60274.MD
also known as H60274-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19916-73-5
IUPAC Name
6-(benzyloxy)-7H-purin-2-amine
Molecular Formula
C12H11N5O
InChI Key
KRWMERLEINMZFT-UHFFFAOYSA-N
SMILES
NC1=NC(OCC2=CC=CC=C2)=C2NC=NC2=N1
Assay (HPLC)
>97.5%

Description

O6-Benzylguanine is an effective substrate and efficient inactivator of DNA alkyltransferase. O6-Benzylguanine is an inhibitor of MGMT. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O6-Benzylguanine is an effective substrate and efficient inactivator of DNA alkyltransferase. O6-Benzylguanine is an inhibitor of MGMT. It is also used as an important raw material and intermediate used in organic Synthesis, pharmac
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text