This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H60364.03
also known as H60364-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material(+)-1,1'-Bis[(2R,5R)-2,5-diethyl-1-phospholanyl]ferrocene
CAS147762-89-8
Melting Point85°C to 91°C
Recommended StorageAmbient temperatures; Store under Argon
TSCANo
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.