It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.
Solubility
Slightly soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
Kazuhiro Kudo.; Noriyoshi Yamamoto. Scalable Synthesis of 8-Amino-3-hydroxy-6H-benzo[c]chromen-6-one: Key Intermediate for SEGRA via the Hurtley Reaction.Org. Process Res. Dev. 2015, 19 (1),309-314.