Thermo Scientific Chemicals

L-Dihydroorotic acid, 98%, Thermo Scientific Chemicals

Catalog number: H60429.03
1 g, Each
Thermo Scientific Chemicals

L-Dihydroorotic acid, 98%, Thermo Scientific Chemicals

Catalog number: H60429.03
1 g, Each
Quantity
Catalog number: H60429.03
also known as H60429-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5988-19-2
IUPAC Name
(4S)-2,6-dioxo-1,3-diazinane-4-carboxylic acid
Molecular Formula
C5H6N2O4
InChI Key
UFIVEPVSAGBUSI-REOHCLBHSA-N
SMILES
OC(=O)[C@@H]1CC(=O)NC(=O)N1
Assay (Titration)
>97.5%

Description

L-Dihydroorotic acid a metabolite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Dihydroorotic acid a metabolite.

Solubility
Soluble in water (partly), and dimethyl formamide.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text