3,4,6-Tri-O-benzyl-D-glucal, is an important building block for both solid- and solution-phase synthesis of oligosaccharides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4,6-Tri-O-benzyl-D-glucal, is an important building block for both solid- and solution-phase synthesis of oligosaccharides.
Solubility
Insoluble in water, easily soluble in CHCl3
Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
- RW Scott.; CH Heathcock. An efficient synthesis of 3, 4, 6-tri-O-benzyl-2-C-methyl-d-glucal. Carbohydrate research. 1996291 (1), 205-208.
- V Bolitt.; C Mioskowski.; SG Lee. Direct preparation of 2-deoxy-D-glucopyranosides from glucals without Ferrier rearrangement. J. Org. Chem. 199055 (23), 5812-5813.