Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF, Thermo Scientific Chemicals
Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H60493.06
also known as H60493-06
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
Add to cart
Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF, Thermo Scientific Chemicals
Catalog numberH60493.06
Price (USD)/ Each
124.65
Online exclusive
138.00 
Save 13.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialBis[2-(di-tert-butylphosphino)ethyl]amine
CAS944710-34-3
Health Hazard 1H225-H302-H315-H318-H335-H336-H351
Health Hazard 2GHS H Statement
H225-H351-H315-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P363-P370+P378q-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
This compound is involved in the transition metal-catalyzed addition of amines to acrylic acid derivatives.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Motoi Kawatsura.; John F. Hartwig. Transition Metal-Catalyzed Addition of Amines to Acrylic Acid Derivatives. A High-Throughput Method for Evaluating Hydroamination of Primary and Secondary Alkylamines.Organometallics. 2001, 20 (10),1960-1964.