This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications This compound is involved in the transition metal-catalyzed addition of amines to acrylic acid derivatives.
Solubility Insoluble in water.
Notes Air Sensitive. Incompatible with oxidizing agents.