Thermo Scientific Chemicals

1,2-Bis(triethoxysilyl)ethane, 95%, Thermo Scientific Chemicals

Catalog number: H60530.AC
25 mL, Each
Thermo Scientific Chemicals

1,2-Bis(triethoxysilyl)ethane, 95%, Thermo Scientific Chemicals

Catalog number: H60530.AC
25 mL, Each
Quantity
Catalog number: H60530.AC
also known as H60530-AC
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16068-37-4
IUPAC Name
4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane
Molecular Formula
C14H34O6Si2
InChI Key
IZRJPHXTEXTLHY-UHFFFAOYSA-N
SMILES
CCO[Si](CC[Si](OCC)(OCC)OCC)(OCC)OCC
Assay (GC)
>94.0%

Description

1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.

Solubility
Soluble in alcohol, aromatic and aliphatic hydrocarbons. Hydrolyzes with water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text