Dibenzenesulfonamide, 95%, Thermo Scientific Chemicals
Dibenzenesulfonamide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dibenzenesulfonamide, 95%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
250 mg
Catalog number H60569.MD
also known as H60569-MD
Price (USD)/ Each
50.90
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
50.90
Add to cart
Dibenzenesulfonamide, 95%, Thermo Scientific Chemicals
Catalog numberH60569.MD
Price (USD)/ Each
50.90
-
Add to cart
Chemical Identifiers
CAS2618-96-4
IUPAC NameN-(benzenesulfonyl)benzenesulfonamide
Molecular FormulaC12H11NO4S2
InChI KeyOVQABVAKPIYHIG-UHFFFAOYSA-N
SMILESO=S(=O)(NS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only