3-Bromoadamantane-1-carboxylic acid, 97%, Thermo Scientific Chemicals
3-Bromoadamantane-1-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromoadamantane-1-carboxylic acid, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Catalog number H60580.03
also known as H60580-03
Price (USD)/ Each
107.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
107.00
Add to cart
3-Bromoadamantane-1-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog numberH60580.03
Price (USD)/ Each
107.00
-
Add to cart
Chemical Identifiers
CAS21816-08-0
IUPAC Name(5R,7S)-3-bromoadamantane-1-carboxylate
Molecular FormulaC11H14BrO2
InChI KeyDJUDQBVINJIMFO-JZVMUCMXSA-M
SMILES[O-]C(=O)C12C[C@H]3C[C@H](CC(Br)(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only