Thermo Scientific Chemicals

Bis(trimethylsiloxy)methylsilane, 97%, Thermo Scientific Chemicals

Catalog number: H60586.AD
50 mL, Each
Thermo Scientific Chemicals

Bis(trimethylsiloxy)methylsilane, 97%, Thermo Scientific Chemicals

Catalog number: H60586.AD
50 mL, Each
Quantity
Catalog number: H60586.AD
also known as H60586-AD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1873-88-7
IUPAC Name
2,2,4,6,6-pentamethyl-3,5-dioxa-2,4,6-trisilaheptan-4-yl
Molecular Formula
C7H21O2Si3
InChI Key
SWGZAKPJNWCPRY-UHFFFAOYSA-N
SMILES
C[Si](O[Si](C)(C)C)O[Si](C)(C)C
Assay (GC)
>96.0%

Description

Bis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(trimethylsiloxy)methylsilane is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

Solubility
Miscible with acetone, ethanol, diethyl ether. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text