Thermo Scientific Chemicals

Diethyl iminodiacetate, 97%, Thermo Scientific Chemicals

Catalog number: H60608.AC
25 mL, Each
Thermo Scientific Chemicals

Diethyl iminodiacetate, 97%, Thermo Scientific Chemicals

Catalog number: H60608.AC
25 mL, Each
Quantity
Catalog number: H60608.AC
also known as H60608-AC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6290-05-7
IUPAC Name
ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
Molecular Formula
C8H15NO4
InChI Key
LJDNMOCAQVXVKY-UHFFFAOYSA-N
SMILES
CCOC(=O)CNCC(=O)OCC
Assay (GC)
>96.0%

Description

It is mainly used for the manufacture of N-(Phosphonomethyl) iminodiacetic acid and glyphosate. Used as raw material for ion exchange resin; for rubbers, electroplating and food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used for the manufacture of N-(Phosphonomethyl) iminodiacetic acid and glyphosate. Used as raw material for ion exchange resin; for rubbers, electroplating and food additives.

Solubility
Moderately soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text