Thermo Scientific Chemicals

4-(4-Methoxyphenyl)butyric acid, 98%, Thermo Scientific Chemicals

Catalog number: H60613.03
1 g, Each
Thermo Scientific Chemicals

4-(4-Methoxyphenyl)butyric acid, 98%, Thermo Scientific Chemicals

Catalog number: H60613.03
1 g, Each
Quantity
Catalog number: H60613.03
also known as H60613-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4521-28-2
IUPAC Name
4-(4-methoxyphenyl)butanoic acid
Molecular Formula
C11H14O3
InChI Key
LZHMNCJMXQKSBY-UHFFFAOYSA-N
SMILES
COC1=CC=C(CCCC(O)=O)C=C1
Appearance (Color)
Cream to pale yellow
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
56-61?C

Description

4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.

Solubility
Slightly soluble in methanol. Insoluble in chloroform.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text