Thermo Scientific Chemicals

1H-Pyrazole-1-carboxamidine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: H60631.18
50 g, Each
Thermo Scientific Chemicals

1H-Pyrazole-1-carboxamidine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: H60631.18
50 g, Each
Quantity
Catalog number: H60631.18
also known as H60631-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4023-02-3
IUPAC Name
1H-pyrazole-1-carboximidamide
Molecular Formula
C4H6N4
InChI Key
UCQFSGCWHRTMGG-UHFFFAOYSA-N
SMILES
NC(=N)N1C=CC=N1
Assay (HPLC)
>98.5%

Description

1H-Pyrazole-1-carboxamidine hydrochloride may be used in the following studies:
  • Preparation of guanidylated hollow fiber membranes.
  • Guanylation of amines and in peptide synthesis.
  • Synthesis of bis-guanidinium-cholesterol derivatives.

    This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

    Applications
    1H-Pyrazole-1-carboxamidine hydrochloride may be used in the following studies: • Preparation of guanidylated hollow fiber membranes. • Guanylation of amines and in peptide synthesis. • Synthesis of bis-guanidinium-cholesterol derivatives.

    Solubility
    Soluble in water.

    Notes
    Incompatible with strong oxidizing agents. Store under room temperature.
    RUO – Research Use Only

    Figures

    Documents & Downloads

    Certificates

      Frequently asked questions (FAQs)

      Citations & References

      Search citations by name, author, journal title or abstract text